Anti-Vaspin Mouse Monoclonal Antibody [clone: VP63]

Lieferant: AdipoGen Life Sciences
AG20A0044C100 AG20A0045C050
ADIPAG20A0044C100EA 390 CHF
ADIPAG20A0044C100 ADIPAG20A0045C050
Anti-Vaspin Mouse Monoclonal Antibody [clone: VP63]
Antikörper
Type:
Antigen: Vaspin
Clonality: Monoclonal
Clone: VP63
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR