Anti-ST2 Mouse Monoclonal Antibody [clone: ST33868]

Lieferant: ADIPOGEN
AG20A0043C100 AG20A0044C050
ADIPAG20A0043C100EA 390 CHF
ADIPAG20A0043C100 ADIPAG20A0044C050
Anti-ST2 Mouse Monoclonal Antibody [clone: ST33868]
Antikörper
Type:
Antigen: ST2
Clonality: Monoclonal
Clone: ST33868
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR