Anti-IL33 Mouse Monoclonal Antibody [clone: IL33026B]

Lieferant: AdipoGen Life Sciences
AG20A0042C100 AG20A0043C050
ADIPAG20A0042C100EA 190 CHF
ADIPAG20A0042C100 ADIPAG20A0043C050
Anti-IL33 Mouse Monoclonal Antibody [clone: IL33026B]
Antikörper
Type:
Antigen: IL33
Clonality: Monoclonal
Clone: IL33026B
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR