Anti-ADIPOQ Mouse Monoclonal Antibody [clone: RADI 264]

Lieferant: ADIPOGEN
AG20A0035C100 AG20A0036C050
ADIPAG20A0035C100EA 350 CHF
ADIPAG20A0035C100 ADIPAG20A0036C050
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: RADI 264]
Antikörper
Type:
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: RADI 264
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR