Anti-Nampt Mouse Monoclonal Antibody [clone: OMNI379]

Lieferant: AdipoGen Life Sciences
AG20A0034BC050 AG20A0034C050
ADIPAG20A0034BC050EA 350 CHF
ADIPAG20A0034BC050 ADIPAG20A0034C050
Anti-Nampt Mouse Monoclonal Antibody [clone: OMNI379]
Antikörper
Type:
Antigen: Nampt
Clonality: Monoclonal
Clone: OMNI379
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR