Anti-RELMB Rat Monoclonal Antibody [clone: MRB 46L]

Lieferant: AdipoGen Life Sciences
AG20A0025C100 AG20A0026C050
ADIPAG20A0025C100EA 360 CHF
ADIPAG20A0025C100 ADIPAG20A0026C050
Anti-RELMB Rat Monoclonal Antibody [clone: MRB 46L]
Antikörper
Type:
Antigen: RELMB
Clonality: Monoclonal
Clone: MRB 46L
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR