Anti-GITR Rat Monoclonal Antibody [clone: AIT 158D]

Lieferant: ADIPOGEN
AG20A0017C050 AG20A0017C100
ADIPAG20A0017C050EA 270 CHF
ADIPAG20A0017C050 ADIPAG20A0017C100
Anti-GITR Rat Monoclonal Antibody [clone: AIT 158D]
Antikörper
Type:
Antigen: GITR
Clonality: Monoclonal
Clone: AIT 158D
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR