Anti-RELMB Mouse Monoclonal Antibody [clone: HRB 149]

Lieferant: AdipoGen Life Sciences
AG20A0012C050 AG20A0012C100
ADIPAG20A0012C050EA 110 CHF
ADIPAG20A0012C050 ADIPAG20A0012C100
Anti-RELMB Mouse Monoclonal Antibody [clone: HRB 149]
Antikörper
Type:
Antigen: RELMB
Clonality: Monoclonal
Clone: HRB 149
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR