Anti-ADIPOQ Mouse Monoclonal Antibody [clone: MADI 1147]

Lieferant: AdipoGen Life Sciences
AG20A0003C050 AG20A0003C100
ADIPAG20A0003C050EA 270 CHF
ADIPAG20A0003C050 ADIPAG20A0003C100
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: MADI 1147]
Antikörper
Type:
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: MADI 1147
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR