Anti-TGFA Mouse Monoclonal Antibody (CF647) [clone: P/T1]

Lieferant: Biotium
BNC470787-500 BNC470787-100
BNC470787-500EA 671 CHF
BNC470787-500 BNC470787-100
Anti-TGFA Mouse Monoclonal Antibody (CF647) [clone: P/T1]
Antikörper
This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: P/T1
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rabbit, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR