Anti-Mitochondrial Marker Mouse Monoclonal Antibody [clone: MTC754]
Lieferant: Biotium
BNUB0754-500 BNUB0754-100
BNUB0754-500EA 613 CHF
BNUB0754-500 BNUB0754-100
Anti-Mitochondrial Marker Mouse Monoclonal Antibody [clone: MTC754]
Antikörper
This MAb recognizes a 60 kDa antigen associated with the mitochondria in cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in cells from a wide variety of animals including insects and bacteria. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignant cells and may be used to stain mitochondria of cells in frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations.
  • Purified
Type: Primary
Antigen: Mitochondrial Marker
Clonality: Monoclonal
Clone: MTC754
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human