Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: MUC1/520]

Lieferant: Biotium
BNC940520-500 BNC940520-100
BNC940520-500EA 671 CHF
BNC940520-500 BNC940520-100
Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: MUC1/520]
Antikörper
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinomas. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: MUC1/520
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR