Anti-CD38 Mouse Monoclonal Antibody (CF647) [clone: AT1]
Lieferant: Biotium
BNC470430-500 BNC470430-100
BNC470430-500EA 671 CHF
BNC470430-500 BNC470430-100
Anti-CD38 Mouse Monoclonal Antibody (CF647) [clone: AT1]
Antikörper
CD38 is a type II transmembrane glycoprotein that is present on early B- and T-cell lineages and activated B- and T-cells but is absent from most mature resting peripheral lymphocytes. CD38 is also found on thymocytes, pre-B cells, germinal center B-cells, mitogen-activated T-cells, monocytes and Ig-secreting plasma cells. CD38 is expressed on CD34 cells. The CD34 CD38- population of hematopoietic stems cells defines the most pluripotent cells (e.g. blast colony forming cells).
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: AT1
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human