Anti-Cytokeratins Mouse Monoclonal Antibody (CF647) [clone: C11]

Lieferant: Biotium
BNC470393-500 BNC470393-100
BNC470393-500EA 671 CHF
BNC470393-500 BNC470393-100
Anti-Cytokeratins Mouse Monoclonal Antibody (CF647) [clone: C11]
Antikörper
This MAb recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. This is a broad-spectrum antibody, which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Type: Primary
Antigen: Cytokeratins
Clonality: Monoclonal
Clone: C11
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Frog, Goat, Guinea Pig, Human, Marmoset, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR