Anti-SILV Mouse Monoclonal Antibody (CF405S) [clone: NKI-beteb]
Lieferant: Biotium
BNC040226-500 BNC040226-100
BNC040226-500EA 671 CHF
BNC040226-500 BNC040226-100
Anti-SILV Mouse Monoclonal Antibody (CF405S) [clone: NKI-beteb]
Antikörper
By immunohistochemistry, this antibody specifically recognizes a protein in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin.This Mab labels formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation.
Type: Primary
Antigen: SILV
Clonality: Monoclonal
Clone: NKI-beteb
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Horse, Human