AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Lieferant: ICN Biomedicals
0219989825 0219989850 0219989880 0219989891
ICNA0219989825EA 174 CHF
ICNA0219989825 ICNA0219989850 ICNA0219989880 ICNA0219989891
AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Proteine und Peptide
AlbumiNZ™ protease-reduced BSA is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality.
  • Off-white to light yellowish to brown powder
  • With ≤5% of moisture content
  • Dissolves in ≤20 min at 20 to 25 °C (20% w/w in water)
AlbumiNZ™ protease-reduced BSA is useful for immunoassays where proteases may degrade proteins or where peroxidase or alkaline phospahatase may hydrolyse substrates.
Order Now
TECHNISCHE DATEN
Protein/Peptid-Name Bovine Serum Albumin
Proteinsynonym BSA|Fraction V|Cohn Fraction V
Environmentally Preferable
Spezies Bovine
Ursprung Serum
CAS Nr. 9048-46-8
Purity >97%
Sorte Immunoassay (EIA) Grade
Form Powder
Lagerbedingungen +4 °C
pH 6,5 - 7,5