Kaliumhexacyanoferrat(III) ≥99%, dunkel rotoranges kristallines Pulver ACS
Lieferant: ICN Biomedicals
Synonyms:
Trikaliumhexacyanoferrat
, Ferricyankalium
, rotes Blutlaugensalz
Potassium ferricyanide is the chemical compound with the formula K3[Fe(CN)6]. This bright red salt contains the octahedrally coordinated [Fe(CN)6]3- ion. It is soluble in water and its solution shows some green-yellow fluorescence.
- Auto Ignition: > 100 °C (Lit.)
- Vapor Pressure: ca. .000001 hPa at 25 °C (Lit.)
Potassium ferricyanide is used as an oxidizing agent to remove silver from negatives and positives, a process called dot etching.
Solubility (10% soln): Yellow with no trace of green or brown
Bitte beachten Research Use Only (RUO).
Formel:
K₃Fe(CN)₆ MW: 329,25 g/mol Siedepunkt: 2735 °C (1013 hPa) (Lit.) Melting Pt: 1539 °C (Lit.) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00011392 CAS-Nummer: 13746-66-2 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
Insoluble Matter | ≤0.005% |
Chloride (Cl) | ≤0.01% |
Sulfates (SO42-) | ≤0.01% |
Ferro Compounds | ≤0.05% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...