Reagents for single parameter colorimeter HI-93728, nitrate
HI93728-03
HANNHI93728-03EA
586
CHF
HANNHI93728-03
Reagents for single parameter colorimeter HI-93728, nitrate
Umwelt-Prüfkits
The HI-93728-01 and HI-93728-03 nitrate reagents are suitable for use with the HI-96728 portable photometer and the HI-83200-02 and HI-83099-02 multiparameter bench photometers.