Serological pipettes, with reservoir
Lieferant: TPP
TPPA94550EA 87.6 CHF
TPPA94550 TPPA394525 TPPA94525 TPPA394550
Serological pipettes, with reservoir
Pipetten Serologische Pipetten
These serological pipettes with reservoir are manufactured from highest grade PS. The reservoir offers large capacity volumes in a compact, ergonomic design. It augments the total volume capacity without adding proportionate length to the pipette.
  • Highly visible, bidirectional graduation incl. over-volume graduation
  • Stable, break-resistant, highly low drip pipette tip
  • Slender, short form enables ergonomic working conditions in the sterile hood
  • Small outside diameter of the tube at the tip resulting in an excellent
  • accessibility of these large pipettes even in small tissue culture flasks
  • Optimised form of the mouthpiece
  • From pyrogens, non cytotoxic and non-haemolytic
  • Fits in pipette aid Turbo-Fix or all commercially available pipette aids with rubber inserts
The triple bagged packaging (3-B) from TPP meets the increasingly more stringent clean room requirements. With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into cleanroom areas. The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
Zertifizierungen: ISO 9001:2008 certified.
Verpackung: Standard individually wrapped in lint free paper / plastic blister. Packed in colour-coded, stackable TPP dispenser box.
Tripple bagged Triple-wrapped in three peel-open bags marked with a security number. The innermost bag 1 is labeled with the product data: product number and description, lot number and EXP. SAL-value: 10⁻⁶ for high sterilisation requirements.