Lithium-tris[(3-ethyl-3-pentyl)oxy]aluminohydride 0.5 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

439111000.EA 121 CHF
439111000.
Lithium-tris[(3-ethyl-3-pentyl)oxy]aluminohydride 0.5 M in THF, AcroSeal®
Lithium-tris[(3-ethyl-3-pentyl)oxy]aluminohydride
Formel: C₂₁H₄₆AlLiO₃
MW: 380,52 g/mol
Dichte: 0,904 g/cm³
Flash-Pt: –17 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012222
CAS-Nummer: 79172-99-9
UN: 3399
ADR: 4.3,I

Order Now

Spezifikation Testergebnisse

Appearance Colorless Solution (May develop some turbidity or precipitate)
Strength 0.40 to 0.60 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR