1,1'-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO437292500EA
200
CHF
ACRO437292500
1,1'-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
1,1'-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate
Formel:
C₃₈H₆₀BF₄FeP₂Rh⁻ MW: 824,4 g/mol Melting Pt: 235…238 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD15144871 CAS-Nummer: 849773-97-3 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
NMR | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...