BippyPhos 97%
Lieferant: Thermo Scientific
ACRO435160010EA
258
CHF
ACRO435160010
BippyPhos 97%
BippyPhos
Formel:
C₃₂H₃₅N₄P MW: 506,634 g/mol Melting Pt: 169…173 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD09038440 CAS-Nummer: 894086-00-1 |
Spezifikation Testergebnisse
Appearance | White to tan Crystalline powder or solid |
Infrared spectrum | Conforms |
NMR | ≥96.0 % |
Specific optical rotation | +64° to +68° (25°C, 589 nm) (c=5.5, CH2Cl2) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...