4-Phenoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

431720500.EA 143 CHF
431720500.
4-Phenoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
4-Phenoxyphenylmagnesium bromide
Formel: C₁₂H₉BrMgO
MW: 273,412 g/mol
Dichte: 0,968 g/cm³
Flash-Pt: –17 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00672009
CAS-Nummer: 21473-02-9
UN: 2924
ADR: 3,II

Order Now

Spezifikation Testergebnisse

Appearance Clear light yellow Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR