Ethylformiat, extra trocken 98+%, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyms:
Ameisensäureethylester
429241000.EA
44.4
CHF
429241000.
Ethylformiat, extra trocken 98+%, AcroSeal®
Ethylformiat
Formel:
C₃H₆O₂ MW: 74,08 g/mol Siedepunkt: 54 °C (760 mmHg) Melting Pt: –80 °C Dichte: 0,917 g/cm³ Flash-Pt: –20 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003294 CAS-Nummer: 109-94-4 UN: 1190 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % |
GC | ≥98.0 % |
UV | 1 cm cell vs HPLC-grade water |
UV | at 240 nm A: ≤0.301 |
UV | at 255 nm A: ≤0.097 |
UV | at 280 nm A: ≤0.015 |
Residue after evaporation | ≤0.0003 % |
Acidity | ≤0.0002 meq/g |
Alkalinity | ≤0.0002 meq/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...