(+)-B-Chlorodiisopinocampheylborane 1.8 M in Hexanen, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
388421000.EA
286
CHF
388421000.
(+)-B-Chlorodiisopinocampheylborane 1.8 M in Hexanen, AcroSeal®
(+)-B-Chlorodiisopinocampheylborane
Formel:
C₂₀H₃₄BCl MW: 320,75 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00074808 CAS-Nummer: 112246-73-8 UN: 2920 ADR: 8,I |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Solution |
Titration Argentometric | 1.65 to 1.85 M |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...