Methyltitanium(IV) triisopropoxide 1 M in THF

Lieferant: Thermo Scientific
Gefahr

ACRO384610500EA 292 CHF
ACRO384610500
Methyltitanium(IV) triisopropoxide 1 M in THF
Methyltitanium(IV) triisopropoxide
Formel: C₁₀H₂₇O₃Ti⁻
MW: 243,19 g/mol
Siedepunkt: 65 °C (760 mmHg)
Dichte: 0,895 g/cm³
Flash-Pt: –17 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD03411233
CAS-Nummer: 18006-13-8
UN: 2056
ADR: 3,II

Order Now

Spezifikation Testergebnisse

Appearance Yellow to orange Solution
Strength 0.9 to 1.1 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR