Propionaldehyd 99+%, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyms: Propanal

427211000.EA 27.4 CHF
427211000.
Propionaldehyd 99+%, AcroSeal®
Propionaldehyd
Formel: C₃H₆O
MW: 58,08 g/mol
Siedepunkt: 48 °C (1013 hPa)
Melting Pt: –81 °C
Dichte: 0,798 g/cm³
Flash-Pt: –40 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00007020
CAS-Nummer: 123-38-6
UN: 1275
ADR: 3,II

Order Now

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3610 to 1.3660 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤20 APHA
Acidity/alkalinity ≤0.00008 meq/g
Substance darkened by H2SO4 passes test
Calcium (Ca) ≤0.2 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Hydrocarbons ≤0.1 % Aromatic hydrocarbons
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤10 ppm
Bromine number ≤0.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR