Boran-Pyridin-Komplex ∼9.4 M 95%, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
427131000.EA
226
CHF
427131000.
Boran-Pyridin-Komplex ∼9.4 M 95%, AcroSeal®
Boranepyridine complex
Appearance: Clear to slightly hazy colorless to amber Liquid
Formel:
C₅H₈BN MW: 92,94 g/mol Melting Pt: 10…11 °C Dichte: 0,92 g/cm³ Flash-Pt: 73 °C Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00012435 CAS-Nummer: 110-51-0 UN: 2810 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | Clear to slightly hazy colorless to amber Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Titration | ≥94 % (Active hydrogen) |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...