4-Fluorobenzylmagnesium chloride 0.25 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

426790500.EA 294 CHF
426790500.
4-Fluorobenzylmagnesium chloride 0.25 M in THF, AcroSeal®
4-Fluorobenzylmagnesium chloride

Appearance: Yellow to orange to brown Solution

Formel: C₇H₆ClFMg
MW: 168,878 g/mol
Dichte: 0,948 g/cm³
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01319898
CAS-Nummer: 1643-73-8
UN: 2056
ADR: 3,II

Order Now

Spezifikation Testergebnisse

Appearance Yellow to orange to brown Solution
Titration Grignard 0.15 to 0.35 M
Refractive index 1.4690 to 1.4730 (20°C, 589 nm)
Tin (Sn) 17.5 to 19.9 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR