L-Selenocystine 98%
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyms: Seleno-L-cystine, (R,R)-3,3'-Diselenobis(2-aminopropionic Acid)
301862500 301860010
ACRO301862500EA 76.4 CHF
ACRO301862500 ACRO301860010
L-Selenocystine 98%
L-Selenocystine
CAS No.: 29621-88-3
Formel: C₆H₁₂N₂O₄Se₂
MW: 334.09 g/mol
Melting Pt: 212…215 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00800971
CAS-Nummer: 29621-88-3
UN: 3283
ADR: 6.1 III
Order Now
Spezifikation Testergebnisse
Appearance Yellow to ochre Powder or crystals
Infrared spectrum Conforms
Titration excess NaOH ≥97.5 %