3-phenylpyrrolidine-2-carboxylic acid 98%

Lieferant: Thermo Scientific
Achtung

ACRO290081000EA 218 CHF
ACRO290081000
3-phenylpyrrolidine-2-carboxylic acid 98%
3-phenylpyrrolidine-2-carboxylic acid

Appearance: White to off-white Powder

Formel: C₁₁H₁₃NO₂
Melting Pt: 234…236 °C
Lager Temperatur: Raumtemperatur
CAS-Nummer: 118758-48-8
UN: 2811
ADR: 6.1,III

Order Now

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +82° to +86° (25°C, 589 nm) (c=0.75, 6 N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR