Antibody extender solution NC, Miser™
Lieferant: Thermo Fisher Scientific
PIER32110EA 292 CHF
PIER32110
Antibody extender solution NC, Miser™
Elektrophorese-Reagenzien Western Blotting Reagents
Miser™ Antibody Extender Solution NC is a post-transfer nitrocellulose membrane-treatment reagent that enables Western blot detection with much less primary antibody.
  • Use at least three times less primary antibody than normal and retain comparable or better signal detectability
  • Easy to perform, 10-minute protocol
  • Works with both colorimetric and chemiluminescent detection modes and with HRP and AP systems