HRP substrate, 1-Step™ Ultra TMB-ELISA
Lieferant: Thermo Fisher Scientific
34028 34029
PIER34028EA 403 CHF
PIER34028 PIER34029
HRP substrate, 1-Step™ Ultra TMB-ELISA
Tests ELISAs
The 1-Step™ Ultra TMB-ELISA contains soluble TMB (3,3´,5,5´-tetramentylbenzidine) that is ideal for sensitive ELISA-based applications. TMB is a chromogenic substrate for horseradish peroxidase. Once oxidised by the enzyme, this substrate yields a blue product that absorbs at 370 nm and 652 nm. Adding sulfuric or phosphoric acid to stop the reaction changes the product to yellow that absorbs at 450 nm and provides greater sensitivity than the blue product. The 1-Step Ultra TMB-ELISA is supplied ready to use with no mixing required.
  • No additional reagents or filtering required
  • Non carcinogenic
  • No DMF or DMSO present
Substrate type: HRP (Horseradish Peroxidase)
Detection method: Colorimeteric method
Order Now
TECHNISCHE DATEN
Environmentally Preferable
Reaktivität der primären Antikörper
Wirt
Learn more