Chromatography cartridges, Protein G, Pierce™
Lieferant: Thermo Fisher Scientific
89926 89927
PIER89926EA 478 CHF
PIER89926 PIER89927
Chromatography cartridges, Protein G, Pierce™
Chromatographiesäulen
Pierce™ Protein G chromatography cartridges are convenient, ready to use prepacked devices for isolation and purification of mammalian polyclonal and monoclonal antibodies from serum, ascites and cell culture supernatants.
  • Immobilised Protein G is ideal for polyclonal IgG purification from mouse, human, cow, goat and sheep serum, including human IgG3 and mouse IgG1 isotypes
  • Resin support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Superior manufacturing method immobilises Protein G by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Binding capacity of 11 to 15 mg human IgG/ml resin
Pierce™ Protein G chromatography cartridges are compatible with the major automated liquid-chromatography systems or manual syringe processing. The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-Lok® syringe fittings or ¹/₁₆" tubing. These cartridges enable fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
Lieferumfang: Kit includes an accessory pack (1 female Luer-Lok® adapter, 1 connector fitting, 1 column plug and bottom caps).