High sensitivity NeutrAvidin™ protein-HRP conjugate, Pierce™
Lieferant: Thermo Fisher Scientific
31030 31032
PIER31030EA 266 CHF
PIER31030 PIER31032
High sensitivity NeutrAvidin™ protein-HRP conjugate, Pierce™
Proteinkonjugate und Markierungen
Pierce™ High Sensitivity NeutrAvidin™-HRP Conjugate is a specially prepared peroxidase conjugated form of avidin biotin binding protein that provides signal amplification like poly-HRP and exceptional storage stability.
  • High specificity, low background binding
  • High sensitivity - detect low levels of target without background; obtain high signal-to-noise ratios
  • Cost effective - uses less conjugate in Western blotting and ELISA applications while still obtaining excellent results
  • Compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates
  • Ready-to-use stabilised liquid format means there is no waiting for thawing and no need to aliquot