Capreomycin sulphate ≥90%

Lieferant: Thermo Scientific
Gefahr

J66684.03EA 241 CHF
J66684.03
Capreomycin sulphate ≥90%
Capreomycin sulphate

Thought to inhibit protein synthesis by inhibiting normal ribosomal functions


  • White powder


Freely soluble in water

Formel: C₂₅H₄₄N₁₄O₈·H₂SO₄
Siedepunkt: 1377 °C
Flash-Pt: >110 °C (230 °F)
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00079032
CAS-Nummer: 1405-37-4
EINECS: 215-776-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR