Chlorimuron ethyl 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
J66605.03
J66605.06
J66605.03EA
58.9
CHF
J66605.03
J66605.06
Chlorimuron ethyl 95%
Chlorimuron ethyl
Purity: 95%
CAS No.: 90982-32-4
Molecular Formula: C15H15ClN4O6S
Formula Weight: 414.82
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Colourless
Melting Point: 180-182°
Solubility: Freely soluble in aqueous solution
MDL No.: MFCD00128063
Formel:
C₁₅H₁₅ClN₄O₆S Melting Pt: 180…182 °C |
MDL Number:
MFCD00128063 CAS-Nummer: 90982-32-4 Merck Index: 14,2093 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...