Cloxacillin sodium salt monohydrate ≥95%

Lieferant: Thermo Scientific
Gefahr

J66500.03EA 133 CHF
J66500.03
Cloxacillin sodium salt monohydrate ≥95%
Cloxacillin sodium salt monohydrate

Applications: Targets primarily gram positive bacteria, especially -lactamase producing Staphylococci

Formel: C₁₉H₁₈ClN₃O₅S
MW: 457,86 g/mol
Siedepunkt: 689 °C
Melting Pt: 170 °C
Flash-Pt: >110 °C (230 °F)
MDL Number: MFCD00150735
CAS-Nummer: 7081-44-9
EINECS: 211-390-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR