Cefdinir 95%
Lieferant: Thermo Fisher Scientific
J66447.03
J66447.06
J66447.03EA
286
CHF
J66447.03
J66447.06
Cefdinir 95%
Cefdinir
Purity: 95%
CAS No.: 91832-40-5
Molecular Formula: C14H13N5O5S2
Formula Weight: 395.41
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 170°
Solubility: Soluble in water
MDL No.: MFCD00865030
Applications: A broad spectrum antibiotic targeting both gram positive and gram negative pathogens
Formel:
C₁₄H₁₃N₅O₅S₂ Melting Pt: 170 °C |
MDL Number:
MFCD00865030 CAS-Nummer: 91832-40-5 |