Meticillin sodium salt ≥99%

Lieferant: Thermo Scientific
Gefahr

J66414.03EA 500 CHF
J66414.03
Meticillin sodium salt ≥99%
Meticillin sodium salt

Applications: Targets primarily the cell wall of gram positive organisms especially Staphylococcus aureus

Formel: C₁₇H₁₉N₂NaO₆S
MW: 402,397 g/mol
Siedepunkt: 640 °C
Melting Pt: 196…197 °C (Zersetzung)
Flash-Pt: >110 °C (230 °F)
MDL Number: MFCD07787409
CAS-Nummer: 132-92-3
EINECS: 205-083-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR