(5E)-5-(4-Ethylbenzyliden)rhodanin

Lieferant: Thermo Scientific
Achtung

J65650.MAEA 176 CHF
J65650.MA
(5E)-5-(4-Ethylbenzyliden)rhodanin
(5E)-5-(4-Ethylbenzyliden)rhodanin

Applications: A c-Myc inhibitor that induces apoptosis

Formel: C₁₂H₁₁NOS₂
MW: 249,35 g/mol
MDL Number: MFCD00758517
CAS-Nummer: 403811-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR