Bolton-Hunter reagent (sulpho-SHPP)

Lieferant: Thermo Scientific
Achtung

Synonyms: Sulfo-SHPP

J64508.03EA 218 CHF
J64508.03
Bolton-Hunter reagent (sulpho-SHPP)
Bolton-Hunter reagent (sulpho-SHPP)

Applications: For iodination of proteins that do not contain tyrosine residues

Formel: C₁₃H₁₃NO₅
MW: 263,249 g/mol
Melting Pt: 132…133 °C
MDL Number: MFCD00005515
CAS-Nummer: 34071-95-9
EINECS: 251-818-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR