DNS Natriumsalz (Desoxyribonukleinsäure Natriumsalz) (aus Kalbsthymus)
Lieferant: Thermo Scientific
J64400.03EA 833 CHF
J64400.03
DNS Natriumsalz (Desoxyribonukleinsäure Natriumsalz) (aus Kalbsthymus)
DNA sodium salt (Deoxyribonucleic acid sodium salt)
Deoxyribonucleic acid sodium salt, is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.
Applications: Suitable for use as a substrate in DNase assays.
Lager Temperatur: Kühlschrank
MDL Number: MFCD00151698
CAS-Nummer: 9007-49-2