Hexokinase
Lieferant: Thermo Fisher Scientific
J63795.4IEA 288 CHF
J63795.4I
Hexokinase
Enzyme
  • Hygroscopic
Applications: Catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP