Thapsigargin 95%
Lieferant: Thermo Scientific
|
Gefahr
|
J62866.MAEA
1610
CHF
J62866.MA
Thapsigargin 95%
Thapsigargin
Cell permeable sesquiterpene lactone tumor promoter. Inhibits endoplasmic reticulum calcium-ATPase
Formel:
C₃₄H₅₀O₁₂ MW: 650,762 g/mol |
MDL Number:
MFCD00083511 CAS-Nummer: 67526-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...