Paraformaldehyd 4% in PBS + Mg + EGTA
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyms: Polyoxymethylene
J62478.AK J62478.AP
J62478.AKEA 179 CHF
J62478.AK J62478.AP
Paraformaldehyd 4% in PBS + Mg + EGTA
Paraformaldehyd
Storage Temperature: 2°C to 8°C Physical Form: Liquid MDL No.: MFCD00133991
Applications: For histology.
Formel: (CH₂O)n
Melting Pt: 123
Flash-Pt: 70
Lager Temperatur: Kühlschrank
MDL Number: MFCD00133991
CAS-Nummer: 30525-89-4
Merck Index: 13,07096