Oxytetracycline 96%
Lieferant: Thermo Fisher Scientific
Purity: 98+%
CAS No.: 79-57-2
Molecular Formula: C22H24N2O9
Formula Weight: 460.43
Physical Form: Powder
Appearance: Pale yellow to tan
Melting Point: 184-185°
Solubility: Sparingly soluble in aqueous solution at 0.6 mg/mL
MDL No.: MFCD00003700
Applications: Impairs mitochondrial protein synthesis
Formel:
C₂₂H₂₄N₂O₉ Melting Pt: 184…185 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003700 CAS-Nummer: 79-57-2 EINECS: 201-212-8 Merck Index: 14,6976 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...