N-Succinimidyl 4-maleimidobutyrate (GMBS)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyms:
GMBS
J61850.MCEA
467
CHF
J61850.MC
N-Succinimidyl 4-maleimidobutyrate (GMBS)
GMBS (N-Succinimidyl 4-maleimidobutyrate)
Applications: A heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity. Useful in enzyme immunoassays
Formel:
C₁₂H₁₂N₂O₆ MW: 280,24 g/mol Melting Pt: 130…132 °C |
MDL Number:
MFCD00036817 CAS-Nummer: 80307-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...