Enalapril

Lieferant: Thermo Scientific

J60750.06EA 542 CHF
J60750.06 J60750.03
Enalapril
Enalapril

Applications: Long-acting ACE inhibitor and antihypertensive

Formel: C₂₀H₂₈N₂O₅
MW: 376,45 g/mol
Melting Pt: 142…147 °C
MDL Number: MFCD00865774
CAS-Nummer: 75847-73-3
Merck Index: 14,3567

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR