2-Chlor-5-methylpyrimidin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64773.06
H64773.14
H64773.06EA
210
CHF
H64773.06
H64773.14
2-Chlor-5-methylpyrimidin 97%
2-Chloro-5-methylpyrimidine
Purity: 97%
CAS No.: 22536-61-4
Molecular Formula: C5H5ClN2
Formula Weight: 128.56
Physical Form: Solid
Melting Point: 89-92°
Beilstein Reference: 108732
MDL No.: MFCD09260903
Formel:
C₅H₅ClN₂ Melting Pt: 89…92 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD09260903 CAS-Nummer: 22536-61-4 |